Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2,6-Dimethylphenyl)-3-thiosemicarbazide

Supplier: MATRIX SCIENTIFIC MS

057046-10G 057046-50G
101912-166EA 1992.31 USD InStock
101912-166 101912-168
4-(2,6-Dimethylphenyl)-3-thiosemicarbazide
4-(2,6-Dimethylphenyl)-3-thiosemicarbazide
Hazard: Irritant


Formula: C₉H₁₃N₃S
Melting Pt: 220…222 °C (dec.)
MDL Number: MFCD00041281
CAS Number: 71058-35-0
Order Now
Specification Test Results
Melting point 220-222°C(d

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty