Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(3-Ethoxyphenyl)propionic acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

057068-1G 057068-5G
101912-242EA 72.31 USD InStock
101912-242 101912-244
3-(3-Ethoxyphenyl)propionic acid ≥98%
3-(3-Ethoxyphenyl)propionic acid
Hazard: Irritant


Formula: C₁₁H₁₄O₃
Melting Pt: 55-57°C
MDL Number: MFCD00016553
CAS Number: 28945-89-3
Order Now
Specification Test Results
Melting point 55-57°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty