Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-cyano-3-phenylpyruvate

Supplier: MATRIX SCIENTIFIC MS

057071-10G 057071-50G
101912-256EA 176.92 USD InStock
101912-256 101912-258
Ethyl-3-cyano-3-phenylpyruvate
Ethyl-3-cyano-3-phenylpyruvate
Hazard: Irritant


Formula: C₁₂H₁₁NO₃
Melting Pt: 128…130 °C
MDL Number: MFCD00019801
CAS Number: 6362-63-6
Order Now
Specification Test Results
Melting point 128-130°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty