Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-fluoropyridine-2-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

066798-500MG 066798-1G 066798-5G
101932-390EA 298.46 USD InStock
101932-390 101932-392 101932-394
Ethyl-3-fluoropyridine-2-carboxylate ≥95%
Ethyl-3-fluoropyridine-2-carboxylate
Hazard: Irritant


Formula: C₈H₈FNO₂
Melting Pt: 74…76 °C
MDL Number: MFCD11616884
CAS Number: 1187732-69-9
Order Now
Specification Test Results
Melting point 74-76°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty