Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-amino-3-(4-chlorophenyl)propanoate hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

066890-500MG 066890-1G 066890-5G
101932-650EA 330.77 USD InStock
101932-650 101932-652 101932-654
Methyl-3-amino-3-(4-chlorophenyl)propanoate hydrochloride ≥95%
Methyl-3-amino-3-(4-chlorophenyl)propanoate hydrochloride
Hazard: Irritant


Formula: C₁₀H₁₃Cl₂NO₂
Melting Pt: 127…129 °C
MDL Number: MFCD06740367
CAS Number: 124082-19-5
Order Now
Specification Test Results
Melting point 127-129°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty