Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-4-(cyanomethyl)piperidine ≥95%

Supplier: MATRIX SCIENTIFIC MS

066894-500MG 066894-1G 066894-5G
101932-668EA 298.46 USD InStock
101932-668 101932-670 101932-672
1-Boc-4-(cyanomethyl)piperidine ≥95%
1-Boc-4-(cyanomethyl)piperidine
Hazard: Irritant


Formula: C₁₂H₂₀N₂O₂
MW: 224.3000 g/mol
Melting Pt: 76…77 °C
MDL Number: MFCD09801019
CAS Number: 256411-39-9
Order Now
Specification Test Results
Melting point 76-77°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty