Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-1H-pyrrolo[2,3-b]pyridine-4-carboxylate ≥97%

Supplier: MATRIX SCIENTIFIC MS

067012-500MG 067012-1G
101932-964EA 384.62 USD InStock
101932-964 101932-966
Ethyl-1H-pyrrolo[2,3-b]pyridine-4-carboxylate ≥97%
Ethyl-1H-pyrrolo[2,3-b]pyridine-4-carboxylate
Hazard: Irritant


Formula: C₁₀H₁₀N₂O₂
Melting Pt: 127…129 °C
MDL Number: MFCD15071133
CAS Number: 1261588-72-0
Order Now
Specification Test Results
Melting point 127-129°C

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty