Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-6-iodo-4-oxo-1,4-dihydro-3-quinolinecarboxylate

Supplier: MATRIX SCIENTIFIC MS

067171-500MG 067171-1G
101933-382EA 230.77 USD InStock
101933-382 101933-384
Ethyl-6-iodo-4-oxo-1,4-dihydro-3-quinolinecarboxylate
Ethyl-6-iodo-4-oxo-1,4-dihydro-3-quinolinecarboxylate
Hazard: Irritant


Formula: C₁₂H₁₀INO₃
Melting Pt: >300 °C
MDL Number: MFCD01912284
CAS Number: 302949-01-5
Order Now
Specification Test Results
Melting point >300°C

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty