Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-cyano-5-methoxy-4-methylbenzoate

Supplier: MATRIX SCIENTIFIC MS

067197-500MG 067197-1G 067197-5G
101933-482EA 261.54 USD InStock
101933-482 101933-484 101933-486
Methyl-2-cyano-5-methoxy-4-methylbenzoate
Methyl-2-cyano-5-methoxy-4-methylbenzoate
Hazard: Irritant


Formula: C₁₁H₁₁NO₃
MW: 205.22 g/mol
Melting Pt: 150…152 °C
MDL Number: MFCD20921584
CAS Number: 1138220-75-3
Order Now
Specification Test Results
Melting point 150-152°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty