Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-N-[2-(4-bromophenyl)-2-oxoethyl]carbamate

Supplier: MATRIX SCIENTIFIC MS

067206-500MG 067206-1G
101933-524EA 403.08 USD InStock
101933-524 101933-526
tert-Butyl-N-[2-(4-bromophenyl)-2-oxoethyl]carbamate
tert-Butyl-N-[2-(4-bromophenyl)-2-oxoethyl]carbamate
Hazard: Irritant


Formula: C₁₃H₁₆BrNO₃
Melting Pt: 96…98 °C
MDL Number: MFCD20921604
CAS Number: 339185-70-5
Order Now
Specification Test Results
Melting point 96-98°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty