Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-fluoro-5-hydroxybenzoic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

070766-500MG 070766-1G 070766-5G
101946-258EA 298.46 USD InStock
101946-258 101946-260 101946-262
2-Bromo-4-fluoro-5-hydroxybenzoic acid ≥95%
2-Bromo-4-fluoro-5-hydroxybenzoic acid
Hazard: Irritant


Formula: C₇H₄BrFO₃
MW: 235.02 g/mol
Melting Pt: 188…190 °C
MDL Number: MFCD21333206
CAS Number: 91659-19-7
Order Now
Specification Test Results
Melting point 188-190°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty