Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Bis(1,1,1,3,3,3-hexafluoro-2-hydroxypropyl)iodobenzene ≥97%

Supplier: MATRIX SCIENTIFIC MS

001121-1G 001121-5G
101781-002EA 53.85 USD InStock
101781-002 101781-004
3,5-Bis(1,1,1,3,3,3-hexafluoro-2-hydroxypropyl)iodobenzene ≥97%
3,5-Bis(1,1,1,3,3,3-hexafluoro-2-hydroxypropyl)iodobenzene
Hazard: Irritant


Formula: C₁₂H₅F₁₂IO₂
Boiling Pt: 115°C/10 Torr
MDL Number: MFCD04038221
CAS Number: 53173-72-1
Order Now
Specification Test Results
Boiling point 80-81°C
ND20 104385

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty