Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perfluoro(methyldecalin) ≥85%, Technical Grade

Supplier: MATRIX SCIENTIFIC MS

001096-25G 001096-100G
101780-944EA 49.23 USD InStock
101780-944 101780-946
Perfluoro(methyldecalin) ≥85%, Technical Grade
Perfluoro(methyldecalin)
TCA listed: T
Hazard: Irritant


Formula: C₁₁F₂₀
Boiling Pt: 160 °C
Melting Pt: –70 °C
Density: 1.96
MDL Number: MFCD00014326
CAS Number: 306-92-3
Order Now
Specification Test Results
Boiling point 160°C
Melting point -70°C
Density 1.96
ND20 1.3195

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty