Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Fluoro-1,1,2-trichloroethan ≥97%

Supplier: MATRIX SCIENTIFIC MS

009178-1G 009178-5G
101796-576EA 90.77 USD InStock
101796-576 101796-578
1-Fluoro-1,1,2-trichloroethan ≥97%
1-Fluoro-1,1,2-trichloroethan
Hazard: Toxic/Ozone Depleter


Formula: C₂H₂Cl₃F
Boiling Pt: 88 °C
Melting Pt: –140 °C
Density: 1.4921
CAS Number: 811-95-0
Order Now
Specification Test Results
Boiling point 88°C
Melting point -104.7°C
Density 1.4921

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty