Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Dimethylsuccinic anhydride ≥98%

Supplier: MATRIX SCIENTIFIC MS

004095-1G 004095-5G 004095-25G
101786-810EA 15.38 USD InStock
101786-810 101786-812 101786-814
2,2-Dimethylsuccinic anhydride ≥98%
2,2-Dimethylsuccinic anhydride
Hazard: Irritant/Moisture Sensitive/Refrigerate


Formula: C₆H₈O₃
Boiling Pt: 122 °C (20 mmHg)
Melting Pt: 29…31 °C
Density: 1.135
MDL Number: MFCD00022608
CAS Number: 17347-61-4
Order Now
Specification Test Results
Boiling point 122°C/20mm
Melting point 29-31°C
Density 1.135

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty