Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Ethoxy)nonafluorobutane

Supplier: MATRIX SCIENTIFIC MS

001119-25G 001119-100G
101780-998EA 24.85 USD InStock
101780-998 101781-000
1-(Ethoxy)nonafluorobutane
1-(Ethoxy)nonafluorobutane
Non-ozone depleting solvent

TCA listed: T
Hazard: Irritant


Formula: C₆H₅F₉O
Boiling Pt: 78°C
Melting Pt: –138 °C
Density: 1.428
Flash Pt: 39°C
MDL Number: MFCD01862062
CAS Number: 163702-05-4
Order Now
Specification Test Results
Boiling point 78°C
Melting point -138°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty