Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dichloro-1,1,1,3-tetrafluoropropane ≥97%

Supplier: MATRIX SCIENTIFIC MS

008907-1G 008907-5G
101795-784EA 73.85 USD InStock
101795-784 101795-786
2,3-Dichloro-1,1,1,3-tetrafluoropropane ≥97%
2,3-Dichloro-1,1,1,3-tetrafluoropropane
Hazard: Ozone Depleter/Irritant


Formula: C₃H₂Cl₂F₄
MW: 184.95 g/mol
Boiling Pt: 70.1 °C
Melting Pt: –98 °C
MDL Number: MFCD04038303
CAS Number: 146916-90-7
Order Now
Specification Test Results
Boiling point 70.1°C
Melting point -98°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty