Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Thienylacetic acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

018341-5G 018341-100G
101805-974EA 35.38 USD InStock
101805-974 101805-978
3-Thienylacetic acid ≥98%
3-Thienylacetic acid
Hazard: Irritant


Formula: C₆H₆O₂S
Boiling Pt: 129 °C (2.5 mmHg)
Melting Pt: 85…86 °C
MDL Number: MFCD00005473
CAS Number: 6964-21-2
UN: 3261
ADR: 8,III
Order Now
Specification Test Results
Boiling point 129°C/2.5mm
Melting point 85-86°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty