Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluorophenylglyoxal hydrate ≥98%

Supplier: MATRIX SCIENTIFIC MS

003872-1G 003872-5G
101786-348EA 18.46 USD InStock
101786-348 101786-350
4-Fluorophenylglyoxal hydrate ≥98%
4-Fluorophenylglyoxal hydrate
Hazard: Irritant


Formula: C₈H₇FO₃
MW: 170.140 g/mol
Boiling Pt: 115 °C (35 mmHg)
Melting Pt: 80…82 °C
MDL Number: MFCD01320770
CAS Number: 7468-86-2
Order Now
Specification Test Results
Boiling point 115°C/35mm
Melting point 80-82°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty