Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Dichlorodecafluorocyclohexane

Supplier: MATRIX SCIENTIFIC MS

005261-1G 005261-5G
101788-552EA 83.08 USD InStock
101788-552 101788-554
1,2-Dichlorodecafluorocyclohexane
1,2-Dichlorodecafluorocyclohexane
Hazard: Irritant


Formula: C₆Cl₂F₁₀
MW: 332.95 g/mol
Boiling Pt: 108 °C
Melting Pt: 32…34 °C
MDL Number: MFCD01862047
CAS Number: 336-14-1
Order Now
Specification Test Results
Boiling point 108°C
Melting point 32-34°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty