Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Trifluoromethyl)benzo[2,1,3]thiadiazole

Supplier: MATRIX SCIENTIFIC MS

005454-250MG 005454-1G
101788-976EA 41.54 USD InStock
101788-976 101788-978
5-(Trifluoromethyl)benzo[2,1,3]thiadiazole
5-(Trifluoromethyl)benzo[2,1,3]thiadiazole
Hazard: Irritant


Formula: C₇H₃F₃N₂S
Boiling Pt: 100 °C (15 mmHg)
Melting Pt: 36…37 °C
MDL Number: MFCD00114584
CAS Number: 17754-05-1
Order Now
Specification Test Results
Boiling point 100°C/15mm
Melting point 36-37°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty