Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl benzoylmalonate ≥95%

Supplier: MATRIX SCIENTIFIC MS

042973-500MG 042973-5G
101858-518EA 375.38 USD InStock
101858-518 101858-522
Diethyl benzoylmalonate ≥95%
Diethyl benzoylmalonate
Hazard: Irritant


Formula: C₁₄H₁₆O₅
MW: 264.28 g/mol
Boiling Pt: 144…149 °C (0.8 m)
Melting Pt: 166 °C
MDL Number: MFCD00059381
CAS Number: 1087-97-4
Order Now
Specification Test Results
Boiling point 144-149°C/0
Melting point 166°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty