Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-1,1,2,2,3,4-Hexafluorocyclobutane ≥97%

Supplier: MATRIX SCIENTIFIC MS

008988-1G 008988-5G
101796-024EA 55.38 USD InStock
101796-024 101796-026
cis-1,1,2,2,3,4-Hexafluorocyclobutane ≥97%
cis-1,1,2,2,3,4-Hexafluorocyclobutane
Hazard: Irritant


Formula: C₄H₂F₆
Boiling Pt: 63 °C
Density: 1.778
MDL Number: MFCD03094205
CAS Number: 22819-47-2
Order Now
Specification Test Results
Boiling point 63°C
Density 1.778
ND20 1.2985

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty