Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,1,2,3-Pentafluoro-2,5-hexadiene ≥97%

Supplier: MATRIX SCIENTIFIC MS

009080-1G 009080-5G
101796-290EA 106.15 USD InStock
101796-290 101796-292
1,1,1,2,3-Pentafluoro-2,5-hexadiene ≥97%
1,1,1,2,3-Pentafluoro-2,5-hexadiene
Hazard: Flammable


Formula: C₆H₅F₅
Boiling Pt: 63 °C
Density: 1.197
MDL Number: MFCD03094436
CAS Number: 1730-22-9
Order Now
Specification Test Results
Boiling point 63°C
Density 1.197
ND20 1.3354

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty