Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,3,3-Tetrafluoropropyl acrylate ≥97%

Supplier: MATRIX SCIENTIFIC MS

003482-5G 003482-25G
101785-766EA 80 USD InStock
101785-766 101785-768
2,2,3,3-Tetrafluoropropyl acrylate ≥97%
2,2,3,3-Tetrafluoropropyl acrylate
Hazard: Irritant/Refrigerate


Formula: C₆H₆F₄O₂
MW: 186.11 g/mol
Boiling Pt: 132…134 °C
Density: 1.348
Flash Pt: 65 °C
CAS Number: 7383-71-3
Order Now
Specification Test Results
Boiling point 132-134°C
Density 1.348
ND20 1.365

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty