Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA)

Supplier: MATRIX SCIENTIFIC MS

004292-5G 004292-25G
101787-052EA 86.15 USD InStock
101787-052 101787-054
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA)
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA)
Hazard: Irritant/Refrigerate


Formula: C₉H₁₈F₃NOSi
MW: 241.33 g/mol
Boiling Pt: 168…170 °C
Density: 1.023
MDL Number: MFCD00009671
CAS Number: 77377-52-7
Order Now
Specification Test Results
Boiling point 168-170°C
Density 1.023
ND20 1.402

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty