Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Thienyl)-1-propanone ≥98%

Supplier: MATRIX SCIENTIFIC MS

004714-25G 004714-100G
101787-718EA 46.15 USD InStock
101787-718 101787-720
1-(2-Thienyl)-1-propanone ≥98%
1-(2-Thienyl)-1-propanone
Hazard: Irritant


Formula: C₇H₈OS
Boiling Pt: 107°C/11mm
Density: 1.126
MDL Number: MFCD00005446
CAS Number: 13679-75-9
Order Now
Specification Test Results
Boiling point 107°C/11mm
Density 1.126
ND20 1.553

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty