Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Pyridylacetonitrile ≥98%

Supplier: MATRIX SCIENTIFIC MS

011167-25G 011167-100G
101800-048EA 215.38 USD InStock
101800-048 101800-050
2-Pyridylacetonitrile ≥98%
2-Pyridylacetonitrile
Hazard: Irritant


Formula: C₇H₆N₂
Boiling Pt: 76…77 °C (2 mmHg)
Melting Pt: 23…25 °C
Density: 1.059
Flash Pt: 93 °C
MDL Number: MFCD00006346
CAS Number: 2739-97-1
UN: 3439
ADR: 6.1,III
Order Now
Specification Test Results
Boiling point 76-77°C/2mm
Density 1.059
ND20 1.525

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty