Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Heptafluoro-2,2,3-trichlorobutane ≥97%

Supplier: MATRIX SCIENTIFIC MS

005901-5G 005901-25G
101789-716EA 20 USD InStock
101789-716 101789-718
Heptafluoro-2,2,3-trichlorobutane ≥97%
Heptafluoro-2,2,3-trichlorobutane
TCA listed: T
Hazard: Irritant


Formula: C₄Cl₃F₇
Boiling Pt: 98 °C
Density: 1.748
MDL Number: MFCD00018065
CAS Number: 335-44-4
Order Now
Specification Test Results
Boiling point 98°C
Density 1.748
ND20 1.353

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty