Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H,1H-Pentadecafluoro-n-octyl acrylate ≥97%

Supplier: MATRIX SCIENTIFIC MS

006251-5G 006251-25G
101790-268EA 38.46 USD InStock
101790-268 101790-270
1H,1H-Pentadecafluoro-n-octyl acrylate ≥97%
1H,1H-Pentadecafluoro-n-octyl acrylate
TCA listed: T
Hazard: Irritant/Refrigerate/Lachrymator


Formula: C₁₁H₅F₁₅O₂
Boiling Pt: 65°C/5mm
Density: 1.403
Flash Pt: 91°C
MDL Number: MFCD00039248
CAS Number: 307-98-2
Order Now
Specification Test Results
Boiling point 65°C/5mm
Density 1.403
ND20 1.3279

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty