Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Butynyl toluene-4-sulfonate ≥98%

Supplier: MATRIX SCIENTIFIC MS

007065-5G 007065-25G
101791-944EA 90.77 USD InStock
101791-944 101791-946
3-Butynyl toluene-4-sulfonate ≥98%
3-Butynyl toluene-4-sulphonate
Hazard: Irritant


Formula: C₁₁H₁₂O₃S
Boiling Pt: 128 °C (0.01 mmHg)
Density: 1.012
MDL Number: MFCD00041687
CAS Number: 23418-85-1
Order Now
Specification Test Results
Boiling point 128°C/0.01m
Density 1.012
ND20 1.522

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty