Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-(benzylamino)propionate ≥97%

Supplier: MATRIX SCIENTIFIC MS

024613-5G 024613-25G
101818-440EA 23.08 USD InStock
101818-440 101818-442
Ethyl-3-(benzylamino)propionate ≥97%
Ethyl-3-(benzylamino)propionate
Hazard: Irritant/Store in Freezer


Formula: C₁₂H₁₇NO₂
Boiling Pt: 133…135 °C (2 mmHg)
Density: 1.03
MDL Number: MFCD00085541
CAS Number: 23583-21-3
Order Now
Specification Test Results
Boiling point 133-135°C/2
Density 1.03
ND20 1.503

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty