Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Thienyl)butyric acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

024654-5G 024654-25G
101818-564EA 67.69 USD InStock
101818-564 101818-566
4-(2-Thienyl)butyric acid ≥98%
4-(2-Thienyl)butyric acid
Hazard: Irritant


Formula: C₈H₁₀O₂S
MW: 170.23 g/mol
Boiling Pt: 122 °C (0.3 mmHg)
Melting Pt: 13…15 °C
Density: 1.184/19 °C
MDL Number: MFCD00005463
CAS Number: 4653-11-6
Order Now
Specification Test Results
Boiling point 122°C/0.3mm
Density 1.169
ND20 1.532

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty