Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2,3-difluoropyridine ≥98%

Supplier: MATRIX SCIENTIFIC MS

024727-5G 024727-25G 024727-100G
101818-738EA 16.92 USD InStock
101818-738 101818-740 101818-742
5-Chloro-2,3-difluoropyridine ≥98%
5-Chloro-2,3-difluoropyridine
Hazard: Irritant


Formula: C5H2ClF2N
Boiling Pt: 135 °C
Density: 1.442
MDL Number: MFCD04039314
CAS Number: 89402-43-7
Order Now
Specification Test Results
Boiling point 135°C
Density 1.442
ND20 1.4738

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty