Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Cyclopentylamino)propionitrile ≥99%

Supplier: MATRIX SCIENTIFIC MS

003198-5G 003198-25G
101785-390EA 80 USD InStock
101785-390 101785-392
3-(Cyclopentylamino)propionitrile ≥99%
3-(Cyclopentylamino)propionitrile
Hazard: Irritant


Formula: C₈H₁₄N₂
Boiling Pt: 115-116°C/7mm
Density: 0.95
MDL Number: MFCD00060286
CAS Number: 1074-63-1
Order Now
Specification Test Results
Boiling point 115-116°C/7
Density 0.95

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty