Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Heptafluoropropyl-1,2,2,2-tetrafluoroethyl ether ≥97%

Supplier: MATRIX SCIENTIFIC MS

006134-5G 006134-25G
101790-152EA 146.15 USD InStock
101790-152 101790-154
Heptafluoropropyl-1,2,2,2-tetrafluoroethyl ether ≥97%
Heptafluoropropyl-1,2,2,2-tetrafluoroethyl ether
TCA listed: T
Hazard: Irritant/Refrigerate


Formula: C₅HF₁₁O
Boiling Pt: 40…42 °C
Density: 1.538
MDL Number: MFCD00042308
CAS Number: 3330-15-2
Order Now
Specification Test Results
Boiling point 40-42°C
Density 1.538

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty