Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl L-2-isothiocyanato-4-(methylthio)butyrate ≥99%

Supplier: MATRIX SCIENTIFIC MS

056601-1G 056601-5G
101911-008EA 69.23 USD InStock
101911-008 101911-010
Methyl L-2-isothiocyanato-4-(methylthio)butyrate ≥99%
Methyl L-2-isothiocyanato-4-(methylthio)butyrate
Hazard: Irritant-Harmful


Formula: C₇H₁₁NO₂S₂
Boiling Pt: 112 °C (1 mmHg)
Density: 1.18
MDL Number: MFCD09025711
CAS Number: 21055-47-0
Order Now
Specification Test Results
Boiling point 112°C/1mm
Density 1.18

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty