Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-isothiocyanatoropionate ≥99%

Supplier: MATRIX SCIENTIFIC MS

056604-1G 056604-5G
101911-018EA 3050.77 USD InStock
101911-018 101911-020
Methyl-3-isothiocyanatoropionate ≥99%
Methyl 3-isothiocyanatoropionate
Hazard: Irritant-Harmful


Formula: C₅H₇NO₂S
Boiling Pt: 40°C/1.5mm
Density: 1.18
MDL Number: MFCD00041129
CAS Number: 18967-35-6
Order Now
Specification Test Results
Boiling point 40°C/1.5mm
Density 1.18

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty