Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloropentyl isothiocyanate ≥98%

Supplier: MATRIX SCIENTIFIC MS

057009-10G 057009-50G
101912-030EA 152.31 USD InStock
101912-030 101912-032
2,5-Dichloropentyl isothiocyanate ≥98%
2,5-Dichloropentyl isothiocyanate
Hazard: Irritant-Harmful


Formula: C₆H₉Cl₂NS
Boiling Pt: 105 °C (1 mmHg)
Density: 1.26
MDL Number: MFCD01310806
CAS Number: 380482-28-0
Order Now
Specification Test Results
Boiling point 105°C/1mm
Density 1.26

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty