Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexafluoro-2-(p-tolyl)isopropanol ≥95%

Supplier: MATRIX SCIENTIFIC MS

004778-5G 004778-25G
101787-800EA 103.08 USD InStock
101787-800 101787-802
Hexafluoro-2-(p-tolyl)isopropanol ≥95%
Hexafluoro-2-(p-tolyl)isopropanol
Hazard: Irritant


Formula: C₁₀H₈F₆O
Boiling Pt: 182°C
MDL Number: MFCD00015357
CAS Number: 2010-61-9
Order Now
Specification Test Results
Boiling point 182°C
ND20 1.4238

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty