Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(4-fluorophenyl)ether ≥97%

Supplier: MATRIX SCIENTIFIC MS

005572-1G 005572-5G 005572-25G
101789-148EA 32.31 USD InStock
101789-148 101789-150 101789-152
Bis(4-fluorophenyl)ether ≥97%
Bis(4-fluorophenyl)ether
Hazard: Irritant


Formula: C₁₂H₈F₂O
MW: 206.19 g/mol
Boiling Pt: 232…240 °C (743 mmHg)
MDL Number: MFCD00013552
CAS Number: 330-93-8
Order Now
Specification Test Results
Boiling point 232-240°C/7
ND20 1.5375

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty