Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,1,2,2,3,3-Heptafluoro-4,6-heptanedione ≥97%

Supplier: MATRIX SCIENTIFIC MS

001165-5G 001165-25G
101781-058EA 127.69 USD InStock
101781-058 101781-060
1,1,1,2,2,3,3-Heptafluoro-4,6-heptanedione ≥97%
1,1,1,2,2,3,3-Heptafluoro-4,6-heptanedione
Hazard: Irritant


Formula: C₇H₅F₇O₂
Boiling Pt: 56…57 °C (38 mmHg)
MDL Number: MFCD00511282
CAS Number: 356-30-9
Order Now
Specification Test Results
Boiling point 56-57°C/38m

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty