Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-methoxy-5-(trifluoromethyl)pyridine ≥97%

Supplier: MATRIX SCIENTIFIC MS

002148-1G 002148-5G
101783-330EA 29.23 USD InStock
101783-330 101783-332
3-Chloro-2-methoxy-5-(trifluoromethyl)pyridine ≥97%
3-Chloro-2-methoxy-5-(trifluoromethyl)pyridine
Hazard: Irritant


Formula: C₇H₅ClF₃NO
MW: 211.57 g/mol
Boiling Pt: 173…174 °C
MDL Number: MFCD00153096
CAS Number: 175136-17-1
Order Now
Specification Test Results
Boiling point 173-174°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty