Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-(Trifluoromethyl)phenoxy)acetonitrile ≥95%

Supplier: MATRIX SCIENTIFIC MS

045222-500MG 045222-1G 045222-5G
101869-766EA 181.54 USD InStock
101869-766 101869-768 101869-770
(3-(Trifluoromethyl)phenoxy)acetonitrile ≥95%
(3-(Trifluoromethyl)phenoxy)acetonitrile
Hazard: Irritant


Formula: C₉H₆F₃NO
Boiling Pt: 82…84 °C (0.5 mmHg)
CAS Number: 2145-31-5
UN: 3276
ADR: 6.1,III
Order Now
Specification Test Results
Boiling point 82-84°C/0.5

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty