Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methylsulfanyl)benzenol ≥95%

Supplier: MATRIX SCIENTIFIC MS

045420-500MG 045420-1G 045420-5G
101870-742EA 173.85 USD InStock
101870-742 101870-744 101870-746
3-(Methylsulfanyl)benzenol ≥95%
3-(Methylsulfanyl)benzenol
Hazard: Irritant


Formula: C₇H₈OS
Boiling Pt: 105…108 °C (2 mmHg)
MDL Number: MFCD11207488
CAS Number: 3463-03-4
Order Now
Specification Test Results
Boiling point 105-108°C/2

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty