Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Methylimidazo[1,2-a]pyridine ≥95%

Supplier: MATRIX SCIENTIFIC MS

045473-500MG 045473-1G 045473-5G
101871-008EA 149.23 USD InStock
101871-008 101871-010 101871-012
8-Methylimidazo[1,2-a]pyridine ≥95%
8-Methylimidazo[1,2-a]pyridine
Hazard: Irritant


Formula: C₈H₈N₂
Boiling Pt: 69°C/0.1mm
MDL Number: MFCD05663805
CAS Number: 874-10-2
Order Now
Specification Test Results
Boiling point 69°C/0.1mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty