Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-methoxytetrafluoropropionate ≥99%

Supplier: MATRIX SCIENTIFIC MS

008786-5G 008786-25G
101795-452EA 50.77 USD InStock
101795-452 101795-454
Methyl-2-methoxytetrafluoropropionate ≥99%
Methyl-2-methoxytetrafluoropropionate
Hazard: Irritant


Formula: C₅H₆F₄O₃
MDL Number: MFCD00155951
CAS Number: 10186-63-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty