Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-chlorotetrafluoropropionate ≥97%

Supplier: MATRIX SCIENTIFIC MS

003982-5G 003982-25G
101786-562EA 118.46 USD InStock
101786-562 101786-564
Ethyl-2-chlorotetrafluoropropionate ≥97%
Ethyl-2-chlorotetrafluoropropionate
Hazard: Irritant


Formula: C₅H₅ClF₄O₂
MW: 208.54 g/mol
Boiling Pt: 104…105 °C
MDL Number: MFCD00155714
CAS Number: 5829-03-8
Order Now
Specification Test Results
Boiling point 104-105°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty