Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Methylenedioxybenzyl chloride 50% (w/w) in methylene chloride , 98%

Supplier: MATRIX SCIENTIFIC MS

004299-5G 004299-25G
101787-060EA 36.92 USD InStock
101787-060 101787-062
3,4-Methylenedioxybenzyl chloride 50% (w/w) in methylene chloride , 98%
3,4-Methylenedioxybenzyl chloride
Hazard: Irritant/Moisture Sensitive/Refrigerate/Lachrymator


Formula: C₈H₇ClO₂
Boiling Pt: 97…100 °C (1 mmHg)
MDL Number: MFCD00044328
CAS Number: 20850-43-5
Order Now
Specification Test Results
Boiling point 97-100°C/1m

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty