Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dibromo-2-chloro-1,1,1,4,4,4-hexafluorobutane ≥97%

Supplier: MATRIX SCIENTIFIC MS

004539-1G 004539-5G
101787-432EA 63.08 USD InStock
101787-432 101787-434
2,3-Dibromo-2-chloro-1,1,1,4,4,4-hexafluorobutane ≥97%
2,3-Dibromo-2-chloro-1,1,1,4,4,4-hexafluorobutane
Hazard: Irritant


Formula: C₄HBr₂ClF₆
Boiling Pt: 78 °C (15 mmHg)
MDL Number: MFCD01862049
CAS Number: 63573-66-0
Order Now
Specification Test Results
Boiling point 78°C/15mm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty